Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005462.4 | NP_005453.2 | 4 | 3557 | ||
XM_011531418.2 | XP_011529720.1 | 4 | 3493 | |||
GenBank mRNA | AF056334.1 | - | 1 | 3519 | ||
BC127771.1 | - | 3 | 807 | |||
BC153867.1 | - | 3559 |