Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206729.1 | NP_001193658.1 | 11 | 1299 | ||
NM_005465.4 | NP_005456.1 | 10 | 1258 | |||
NM_181690.2 | NP_859029.1 | 10 | 1258 | |||
XM_005272994.4 | XP_005273051.1 | 11 | 1483 | |||
XM_005272995.2 | XP_005273052.1 | 11 | 1289 | |||
XM_011544012.2 | XP_011542314.1 | 11 | 1483 | |||
XM_011544014.2 | XP_011542316.1 | 4 | 666 | |||
XM_016999985.1 | XP_016855474.1 | 12 | 1431 | |||
GenBank mRNA | AF085234.1 | - | 10 | 1193 | ||
AF124141.1 | - | 10 | 1158 | |||
AF135794.1 | - | 10 | 1146 | |||
AJ245709.1 | - | 10 | 1156 | |||
AK308052.1 | - | 10 | 1184 | |||
AL117525.1 | - | 11 | 1299 | |||
AY005799.1 | - | 10 | 1219 | |||
BC020479.1 | - | 10 | 1192 | |||
BC121154.2 | - | 10 | 1229 | 40121374 |