Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286121.1 | NP_001273050.1 | 8 | 958 | ||
NM_005495.2 | NP_005486.1 | 7 | 858 | |||
XM_011514211.1 | XP_011512513.1 | 7 | 897 | |||
XM_011514213.2 | XP_011512515.1 | 6 | 650 | |||
XM_011514214.2 | XP_011512516.1 | 7 | 835 | |||
XM_011514215.2 | XP_011512517.1 | 7 | 835 | |||
XM_011514218.2 | XP_011512520.1 | 3 | 186 | |||
XM_017010150.1 | XP_016865639.1 | 7 | 835 | |||
GenBank mRNA | AB020527.1 | - | 7 | 847 | ||
AK024903.1 | - | 7 | 858 | |||
BC109208.1 | - | 7 | 886 | 40006623 |