Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042581.1 | NP_001036046.1 | 4 | 670 | ||
NM_001042588.1 | NP_001036053.1 | 4 | 475 | |||
NM_005701.3 | NP_005692.1 | 4 | 775 | |||
GenBank mRNA | AF039029.1 | - | 4 | 420 | ||
AK054641.1 | - | 5 | 908 | |||
AK289475.1 | - | 4 | 471 | |||
BC004203.2 | - | 4 | 406 | 3605328 | ||
BU945130.1 | - | 4 | 569 | |||
CD244425.1 | - | 4 | 670 | |||
CR456811.1 | - | 3 | 330 |