Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015322.4 | NP_056137.1 | 2 | 1764 | ||
XM_017021859.1 | XP_016877348.1 | 2 | 1277 | |||
GenBank mRNA | AB007856.2 | - | 2 | 1149 | ||
AF178632.1 | - | 2 | 1149 | |||
AF204883.1 | - | 2 | 1149 | |||
AK098620.1 | - | 1 | 285 | |||
AK290167.1 | - | 2 | 1498 | |||
AL833264.1 | - | 1 | 2096 | |||
BC010122.2 | - | 2 | 1764 | 3840453 | ||
BC014558.2 | - | 2 | 1764 | 3840377 |