Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286045.1 | NP_001272974.1 | 2 | 324 | ||
NM_001286046.1 | NP_001272975.1 | 2 | 324 | |||
NM_016090.3 | NP_057174.1 | 2 | 324 | |||
GenBank mRNA | AB209753.1 | - | 2 | 222 | ||
AF156098.1 | - | 2 | 202 | |||
AK002015.1 | - | 2 | 237 | |||
AK225615.1 | - | 2 | 190 | |||
AK225667.1 | - | 2 | 215 | |||
AK312619.1 | - | 2 | 280 | |||
BC034381.1 | - | 2 | 191 | 5180771 | ||
BC070262.1 | - | 2 | 185 | 4619267 | ||
DA027521.1 | - | 2 | 324 |