Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005777.2 | NP_005768.1 | 5 | 1688 | ||
XM_005264784.1 | XP_005264841.1 | 5 | 1963 | |||
XM_005264785.1 | XP_005264842.1 | 4 | 379 | |||
XM_005264786.1 | XP_005264843.1 | 4 | 351 | |||
XM_005264787.2 | XP_005264844.1 | 3 | 1117 | |||
XM_006712916.1 | XP_006712979.1 | 5 | 1788 | |||
XM_017005496.1 | XP_016860985.1 | 3 | 1117 | |||
XM_017005499.1 | XP_016860988.1 | 5 | 1963 | |||
XM_017005500.1 | XP_016860989.1 | 5 | 1963 | |||
XM_017005501.1 | XP_016860990.1 | 5 | 1963 | |||
GenBank mRNA | AF042857.1 | - | 5 | 1541 | ||
AF069517.1 | - | 5 | 1644 | |||
AF091264.1 | - | 5 | 1573 | |||
AK093836.1 | - | 4 | 294 | |||
AK291719.1 | - | 5 | 1688 | |||
AK303371.1 | - | 5 | 1581 | |||
AK308794.1 | - | 4 | 303 | |||
BX648535.1 | - | 5 | 1865 | |||
U50839.1 | - | 4 | 1474 |