Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005777.2 | NP_005768.1 | 3 | 666 | ||
XM_005264784.1 | XP_005264841.1 | 3 | 941 | |||
XM_005264787.2 | XP_005264844.1 | 1 | 95 | |||
XM_006712916.1 | XP_006712979.1 | 3 | 766 | |||
XM_017005496.1 | XP_016860985.1 | 1 | 95 | |||
XM_017005499.1 | XP_016860988.1 | 3 | 941 | |||
XM_017005500.1 | XP_016860989.1 | 3 | 941 | |||
XM_017005501.1 | XP_016860990.1 | 3 | 941 | |||
GenBank mRNA | AF042857.1 | - | 3 | 519 | ||
AF069517.1 | - | 3 | 622 | |||
AF091264.1 | - | 3 | 551 | |||
AK291719.1 | - | 3 | 666 | |||
AK303371.1 | - | 3 | 559 | |||
BX648535.1 | - | 3 | 843 | |||
U50839.1 | - | 2 | 452 |