Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006941.2 | NP_001006942.1 | 7, 8 | 1118 | ||
NM_005787.5 | NP_005778.1 | 7, 8 | 1045 | |||
NR_024533.1 | 6, 7 | 945 | ||||
NR_024534.1 | 7, 8 | 1008 | ||||
XM_011512322.1 | XP_011510624.1 | 7, 8 | 930 | |||
XM_011512323.2 | XP_011510625.1 | 7, 8 | 916 | |||
GenBank mRNA | AK289361.1 | - | 7, 8 | 1074 | ||
AK299575.1 | - | 6, 7 | 936 | |||
BC002839.2 | - | 7, 8 | 1020 | 3636604 | ||
BC004313.1 | - | 7, 8 | 1000 | |||
BC021062.1 | - | 7, 8 | 1009 | |||
CX761434.1 | - | 2, 3 | 513 | |||
KP189279.1 | - | 7, 8 | 1038 | |||
KP189280.1 | - | 7, 8 | 1001 | |||
KP189281.1 | - | 6, 7 | 854 | |||
KP189282.1 | - | 7, 8 | 1155 | |||
KP189283.1 | - | 6, 7 | 938 | |||
KP189284.1 | - | 7, 8 | 1036 | |||
KP189285.1 | - | 7, 8 | 999 | |||
KP189286.1 | - | 6, 7 | 854 | |||
KP189287.1 | - | 7, 8 | 954 | |||
KP189288.1 | - | 6, 7 | 936 | |||
KP189289.1 | - | 7, 8 | 1035 | |||
KP189290.1 | - | 7, 8 | 999 | |||
KP189291.1 | - | 6, 7 | 854 | |||
KP189292.1 | - | 7, 8 | 954 | |||
KP189293.1 | - | 6, 7 | 936 | |||
KP189294.1 | - | 7, 8 | 1014 | |||
KP189295.1 | - | 6, 7 | 808 | |||
KP189296.1 | - | 7, 8 | 977 | |||
KP189297.1 | - | 6, 7 | 771 | |||
KP189298.1 | - | 4, 5 | 567 | |||
KP189299.1 | - | 7, 8 | 870 | |||
KP189300.1 | - | 6, 7 | 914 | |||
KP189302.1 | - | 7, 8 | 1035 | |||
KP189303.1 | - | 7, 8 | 997 | |||
KP189304.1 | - | 6, 7 | 854 | |||
KP189305.1 | - | 7, 8 | 954 | |||
KP189306.1 | - | 6, 7 | 934 | |||
KP189307.1 | - | 7, 8 | 1034 | |||
KP189308.1 | - | 7, 8 | 997 | |||
KP189309.1 | - | 6, 7 | 854 | |||
KP189310.1 | - | 7, 8 | 954 | |||
KP189311.1 | - | 6, 7 | 934 | |||
KP189312.1 | - | 7, 8 | 1034 | |||
KP189313.1 | - | 6, 7 | 828 | |||
KP189315.1 | - | 7, 8 | 997 | |||
KP189316.1 | - | 6, 7 | 791 | |||
KP189318.1 | - | 6, 7 | 854 | |||
KP189319.1 | - | 5, 6 | 648 | |||
KP189321.1 | - | 7, 8 | 954 | |||
KP189322.1 | - | 6, 7 | 748 | |||
KP189324.1 | - | 6, 7 | 934 | |||
KP189325.1 | - | 5, 6 | 728 | |||
Y09022.1 | - | 7, 8 | 1045 |