Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005792.2 | NP_005783.2 | 4 | 323 | ||
XM_011522808.2 | XP_011521110.1 | 5 | 603 | |||
XM_017022852.1 | XP_016878341.1 | 4 | 437 | |||
GenBank mRNA | AK222667.1 | - | 4 | 323 | ||
AK314163.1 | - | 4 | 323 | |||
AW175781.1 | - | 4 | 355 | |||
BC005242.1 | - | 4 | 294 | 3997566 | ||
BC011020.1 | - | 4 | 300 | 4287267 | ||
BC031017.1 | - | 4 | 295 | 4670801 | ||
BU595100.1 | - | 4 | 319 | |||
X98263.1 | - | 4 | 306 |