Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164189.1 | NP_001157661.1 | 7 | 1039 | ||
NM_001164194.1 | NP_001157666.1 | 7 | 1003 | |||
NM_005817.4 | NP_005808.3 | 7 | 1039 | |||
GenBank mRNA | AF051314.1 | - | 3 | 351 | ||
AF051315.1 | - | 3 | 327 | |||
AF055574.1 | - | 7 | 915 | |||
AF057140.1 | - | 7 | 936 | |||
AK122671.1 | - | 7 | 890 | |||
AK223054.1 | - | 7 | 969 | |||
AK225045.1 | - | 7 | 905 | |||
AK225400.1 | - | 7 | 920 | |||
AK226088.1 | - | 7 | 917 | |||
AK291104.1 | - | 7 | 996 | |||
AK312740.1 | - | 7 | 965 | |||
BC001590.2 | - | 7 | 912 | 2987965 | ||
BC005818.2 | - | 7 | 928 | 3833411 | ||
BC007566.1 | - | 7 | 928 | 3028104 | ||
BC019278.1 | - | 7 | 928 | 2905275 | ||
BT007235.1 | - | 6 | 862 |