Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177355.2 | NP_001170826.1 | 12 | 1046 | ||
NM_005823.5 | NP_005814.2 | 12 | 1106 | |||
NM_013404.4 | NP_037536.2 | 11 | 1049 | |||
XM_005255034.4 | XP_005255091.1 | 11 | 1352 | |||
XM_011522348.2 | XP_011520650.1 | 12 | 1119 | |||
XM_017022853.1 | XP_016878342.1 | 11 | 1266 | |||
XM_017022854.1 | XP_016878343.1 | 11 | 1277 | |||
XM_017022855.1 | XP_016878344.1 | 11 | 1199 | |||
XM_017022856.1 | XP_016878345.1 | 11 | 1200 | |||
XM_017022857.1 | XP_016878346.1 | 13 | 1196 | |||
XM_017022858.1 | XP_016878347.1 | 12 | 1116 | |||
GenBank mRNA | AF180951.1 | - | 1 | 50 | ||
AY743922.1 | - | 12 | 1106 | |||
BC003512.1 | - | 11 | 1352 | 3611296 | ||
BC009272.2 | - | 12 | 1045 | 3957372 | ||
D49441.1 | - | 11 | 1030 | |||
U40434.1 | - | 11 | 1031 |