Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABEPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174152.1 | NP_001167623.1 | 5 | 515 | ||
NM_005833.3 | NP_005824.2 | 4 | 676 | |||
XM_005251640.4 | XP_005251697.1 | 4 | 588 | |||
XM_005251642.4 | XP_005251699.1 | 4 | 596 | |||
XM_011518120.2 | XP_011516422.1 | 4 | 591 | |||
XM_017014177.1 | XP_016869666.1 | 5 | 500 | |||
XM_017014178.1 | XP_016869667.1 | 5 | 564 | |||
XM_017014179.1 | XP_016869668.1 | 3 | 475 | |||
GenBank mRNA | AK290768.1 | - | 4 | 520 | ||
AL832249.1 | - | 5 | 512 | |||
BC000503.2 | - | 4 | 653 | 2821471 | ||
BC053541.1 | - | 4 | 485 | 5724458 | ||
BC065725.1 | - | 5 | 518 | 4102355 | ||
CR456837.1 | - | 3 | 346 | |||
CR541851.1 | - | 3 | 346 | |||
KU178507.1 | - | 3 | 346 | |||
KU178508.1 | - | 2 | 188 | |||
Z97074.1 | - | 5 | 495 |