Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RABEPK SIRNA Assays ›
Gene Symbol:
RABEPK
Gene Name:
Rab9 effector protein with kelch motifs
Gene Aliases:
RAB9P40, bA65N13.1, p40
Chromosome Location:
Chr.9: 125200493 - 125234159 on Build GRCh38
UniGene ID:
Hs.19012
Species:
Human
siRNA ID
HSS115644
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RABEPK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174152.1
NP_001167623.1
7, 8
838
NM_001174153.1
NP_001167624.1
6, 7
685
NM_005833.3
NP_005824.2
6, 7
999
XM_005251640.4
XP_005251697.1
6, 7
911
XM_005251641.4
XP_005251698.1
5, 6
766
XM_005251642.4
XP_005251699.1
5, 6
757
XM_005251644.4
XP_005251701.1
4, 5
607
XM_011518120.2
XP_011516422.1
6, 7
914
XM_017014177.1
XP_016869666.1
7, 8
823
XM_017014178.1
XP_016869667.1
6, 7
725
XM_017014179.1
XP_016869668.1
5, 6
798
XM_017014180.1
XP_016869669.1
5, 6
572
XM_017014181.1
XP_016869670.1
4, 5
683
XM_017014182.1
XP_016869671.1
5, 6
508
XM_017014183.1
XP_016869672.1
5, 6
845
GenBank mRNA
AK290768.1
-
6, 7
843
BC000503.2
-
6, 7
976
2821471
BC047023.1
-
6, 7
665
5396980
BC053541.1
-
6, 7
808
5724458
BC065725.1
-
7, 8
841
4102355
CR456837.1
-
5, 6
669
CR541851.1
-
5, 6
669
KU178507.1
-
5, 6
669
KU178508.1
-
5, 6
582
KU178509.1
-
4, 5
520
KU178510.1
-
4, 5
516
KU178511.1
-
3, 4
354
Z97074.1
-
7, 8
818
Back To Top
More Information
Gene Ontology Categories:
Function(s)
receptor-mediated endocytosis
vesicle docking involved in exocytosis
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0