Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005850.4 | NP_005841.1 | 3 | 984 | ||
GenBank mRNA | AK097315.1 | - | 3 | 1076 | ||
AK223322.1 | - | 3 | 500 | |||
AK310495.1 | - | 3 | 500 | |||
BC004273.1 | - | 3 | 499 | 3615080 | ||
BC013886.2 | - | 3 | 499 | 3829990 | ||
BC090883.1 | - | 4 | 1104 | 30386953 | ||
BM460550.1 | - | 3 | 542 |
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK302154.1 | - | 5 | 594 |