Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005854.2 | NP_005845.2 | 4 | 442 | ||
GenBank mRNA | AJ001015.1 | - | 4 | 442 | ||
AL548945.3 | - | 4 | 448 | |||
AY265458.1 | - | 4 | 374 | |||
BC027975.1 | - | 4 | 391 | 5229167 | ||
EF687002.1 | - | 4 | 393 |