Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278650.1 | NP_001265579.1 | 3 | 1177 | ||
NM_006057.2 | NP_006048.1 | 3 | 1329 | |||
NM_033170.2 | NP_149360.1 | 3 | 1196 | |||
NM_033171.2 | NP_149361.1 | 4 | 1438 | |||
NM_033172.2 | NP_149362.2 | 3 | 1278 | |||
XM_017028227.1 | XP_016883716.1 | 4 | 2754 | |||
XM_017028228.1 | XP_016883717.1 | 5 | 5378 | |||
XM_017028229.1 | XP_016883718.1 | 5 | 2916 | |||
XM_017028230.1 | XP_016883719.1 | 5 | 2931 | |||
XM_017028231.1 | XP_016883720.1 | 6 | 5487 | |||
XM_017028232.1 | XP_016883721.1 | 6 | 2974 | |||
XM_017028233.1 | XP_016883722.1 | 4 | 2751 | |||
XM_017028234.1 | XP_016883723.1 | 5 | 2913 | |||
XM_017028235.1 | XP_016883724.1 | 4 | 1354 | |||
XM_017028236.1 | XP_016883725.1 | 5 | 3312 | |||
XM_017028237.1 | XP_016883726.1 | 4 | 3427 | |||
XM_017028238.1 | XP_016883727.1 | 5 | 5245 | |||
XM_017028239.1 | XP_016883728.1 | 6 | 5354 | |||
XM_017028240.1 | XP_016883729.1 | 3 | 2523 | |||
XM_017028241.1 | XP_016883730.1 | 3 | 2520 | |||
GenBank mRNA | AB020337.1 | - | 3 | 1322 | ||
AK292951.1 | - | 3 | 1177 | |||
AY372061.1 | - | 5 | 1481 | |||
BC104862.1 | - | 1 | 980 | 8143865 | ||
BC104864.1 | - | 1 | 980 | 8143867 |