Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014254.2 | NP_055069.1 | 1 | 182 | ||
XM_005268562.3 | XP_005268619.1 | 1 | 128 | |||
GenBank mRNA | AB015633.1 | - | 1 | 128 | ||
AK289382.1 | - | 1 | 76 | |||
BC002596.2 | - | 1 | 113 | |||
BC013152.1 | - | 1 | 114 | 3919181 |