Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199573.1 | NP_001186502.1 | 8 | 1300 | ||
NM_006074.4 | NP_006065.2 | 8 | 1312 | |||
GenBank mRNA | AK290434.1 | - | 8 | 1042 | ||
AK298715.1 | - | 9 | 992 | |||
AK298934.1 | - | 8 | 866 | |||
AK301192.1 | - | 7 | 783 | |||
AK310098.1 | - | 7 | 796 | |||
AL360134.1 | - | 6 | 470 | |||
AM040752.1 | - | 5 | 260 | |||
BC022281.1 | - | 8 | 1119 | 4718024 | ||
BC035582.1 | - | 8 | 1085 | 5583800 | ||
HQ842635.1 | - | 7 | 933 | |||
X82200.1 | - | 8 | 1043 |