Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014258.3 | NP_055073.2 | 39 | 4332 | ||
XM_011528487.2 | XP_011526789.1 | 39 | 4368 | |||
XM_011528488.2 | XP_011526790.1 | 39 | 4320 | |||
XM_011528489.2 | XP_011526791.1 | 39 | 4363 | |||
XM_011528490.2 | XP_011526792.1 | 39 | 4344 | |||
XM_011528493.2 | XP_011526795.1 | 38 | 4283 | |||
XM_017027586.1 | XP_016883075.1 | 38 | 4564 | |||
XM_017027587.1 | XP_016883076.1 | 38 | 4561 | |||
XM_017027588.1 | XP_016883077.1 | 37 | 4409 | |||
XM_017027589.1 | XP_016883078.1 | 37 | 4394 | |||
XM_017027590.1 | XP_016883079.1 | 38 | 4257 | |||
XM_017027591.1 | XP_016883080.1 | 30 | 3663 | |||
GenBank mRNA | BC132864.1 | - | 38 | 4201 | 40146886 | |
BC132870.1 | - | 38 | 4203 | 40146892 | ||
BX648091.1 | - | 39 | 4285 | |||
BX649108.1 | - | 39 | 4310 | |||
Y08982.1 | - | 37 | 4111 |