Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098531.2 | NP_001092001.1 | 19 | 2434 | ||
NM_001098532.2 | NP_001092002.1 | 18 | 2198 | |||
NM_006105.5 | NP_006096.2 | 19 | 1968 | |||
XM_005268571.3 | XP_005268628.1 | 18 | 1944 | |||
XM_011537752.1 | XP_011536054.1 | 19 | 2451 | |||
XM_011537755.1 | XP_011536057.1 | 19 | 2013 | |||
XM_011537758.2 | XP_011536060.1 | 19 | 2063 | |||
GenBank mRNA | AF103905.1 | - | 18 | 1758 | ||
AK055502.1 | - | 1 | 749 | |||
AK074444.1 | - | 3 | 1126 | |||
AK092448.1 | - | 19 | 2406 | |||
AK125545.1 | - | 15 | 1451 | |||
AK290230.1 | - | 20 | 2099 | |||
AK299029.1 | - | 18 | 1915 | |||
BC017728.1 | - | 19 | 2048 | 4414360 | ||
BC092404.1 | - | 19 | 2037 | 6181000 | ||
U78168.1 | - | 19 | 1973 |
Gene Symbol: LOC105369748 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL831948.1 | - | 4 | 2211 |