Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006324.2 | NP_006315.1 | 7 | 1006 | ||
GenBank mRNA | AB009285.1 | - | 7 | 978 | ||
AK226031.1 | - | 8 | 994 | |||
AK313948.1 | - | 7 | 948 | |||
BC000991.2 | - | 7 | 1002 | 3449836 | ||
BT009819.1 | - | 7 | 869 | |||
CR542111.1 | - | 7 | 869 | |||
D86549.1 | - | 6 | 805 |