Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006335.2 | NP_006326.1 | 5 | 429 | ||
GenBank mRNA | AF106622.1 | - | 5 | 400 | ||
AK023063.1 | - | 2 | 1603 | |||
AK315602.1 | - | 5 | 417 | |||
BC000294.2 | - | 5 | 399 | 2819455 | ||
BC004439.1 | - | 5 | 399 | 2819455 | ||
BC007106.1 | - | 5 | 410 | 4284092 | ||
BC009784.1 | - | 5 | 421 | 3683125 | ||
BC013823.1 | - | 5 | 388 | |||
BC015098.1 | - | 5 | 427 | 3686369 | ||
BC016817.1 | - | 3 | 284 | |||
BC020833.1 | - | 5 | 407 | 4773149 | ||
X97544.1 | - | 5 | 406 |