Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TACC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006342.2 | NP_006333.1 | 15, 16 | 2598 | ||
XM_005247929.2 | XP_005247986.1 | 15, 16 | 2608 | |||
XM_005247930.2 | XP_005247987.1 | 14, 15 | 1637 | |||
XM_017007653.1 | XP_016863142.1 | 15, 16 | 2650 | |||
GenBank mRNA | AA682436.1 | - | 1, 2 | 247 | ||
AF093543.1 | - | 15, 16 | 2551 | |||
AF479620.1 | - | 14, 15 | 1464 | |||
AI684124.1 | - | 1, 2 | 254 | |||
AJ243997.1 | - | 15, 16 | 2543 | |||
BC106071.1 | - | 15, 16 | 2559 | 6148176 | ||
BC111771.1 | - | 15, 16 | 2511 | 40034847 |