Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006408.3 | NP_006399.1 | 3 | 347 | ||
XM_005249581.4 | XP_005249638.1 | 3 | 316 | |||
GenBank mRNA | AF007791.1 | - | 2 | 170 | ||
AF038451.1 | - | 3 | 218 | |||
AF087879.1 | - | 3 | 309 | |||
AF088867.1 | - | 3 | 307 | |||
AF115926.1 | - | 3 | 217 | |||
AK223067.1 | - | 3 | 238 | |||
AK296721.1 | - | 2 | 502 | |||
AK312942.1 | - | 3 | 228 | |||
AY359009.1 | - | 3 | 307 | |||
BC015503.1 | - | 3 | 216 | 3852448 | ||
BF680655.1 | - | 3 | 348 | |||
BT007048.1 | - | 2 | 160 | |||
FJ224307.1 | - | 2 | 170 | |||
KJ767789.1 | - | 3 | 321 | |||
KJ767793.1 | - | 2, 3 | 160 |