Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168398.1 | NP_001161870.1 | 6 | 669 | ||
NM_006416.4 | NP_006407.1 | 7 | 846 | |||
GenBank mRNA | AJ810303.1 | - | 7 | 652 | ||
AJ851888.1 | - | 6 | 590 | |||
AJ851889.1 | - | 5 | 523 | |||
AJ851890.1 | - | 6 | 477 | |||
AK293696.1 | - | 6 | 638 | |||
AL122119.1 | - | 6 | 774 | |||
BC017807.1 | - | 7 | 794 | 4690513 | ||
D87969.1 | - | 7 | 794 |