Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POSTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135935.1 | NP_001129407.1 | 17 | 2166 | ||
NM_001286665.1 | NP_001273594.1 | 17 | 2166 | |||
NM_006475.2 | NP_006466.2 | 18 | 2247 | |||
XM_005266231.2 | XP_005266288.1 | 18 | 2242 | |||
XM_017020355.1 | XP_016875844.1 | 18 | 2242 | |||
XM_017020356.1 | XP_016875845.1 | 18 | 2242 | |||
GenBank mRNA | AK304640.1 | - | 7 | 535 | ||
AY140646.1 | - | 17 | 2059 | |||
D13666.1 | - | 18 | 2140 | |||
EU262883.1 | - | 17 | 2048 | |||
EU262885.1 | - | 17 | 2048 | |||
GU354210.1 | - | 17 | 2059 |