Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CELF2 SIRNA Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP, Elav-like family member 2
Gene Aliases:
BRUNOL3, CELF-2, CUG-BP2, CUGBP2, ETR-3, ETR3, NAPOR
Chromosome Location:
Chr.10: 10462515 - 11336675 on Build GRCh38
UniGene ID:
Hs.309288
Species:
Human
siRNA ID
HSS116450
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001025076.2
NP_001020247.1
3
540
NM_001025077.2
NP_001020248.1
3
397
NM_001083591.1
NP_001077060.1
3
540
NM_001326317.1
NP_001313246.1
5
399
NM_001326318.1
NP_001313247.1
5
399
NM_001326319.1
NP_001313248.1
6
437
NM_001326320.1
NP_001313249.1
6
493
NM_001326321.1
NP_001313250.1
5
399
NM_001326323.1
NP_001313252.1
6
493
NM_001326324.1
NP_001313253.1
4
376
NM_001326325.1
NP_001313254.1
5
470
NM_001326326.1
NP_001313255.1
4
395
NM_001326327.1
NP_001313256.1
4
395
NM_001326328.1
NP_001313257.1
5
489
NM_001326329.1
NP_001313258.1
3
429
NM_001326330.1
NP_001313259.1
3
429
NM_001326331.1
NP_001313260.1
3
581
NM_001326332.1
NP_001313261.1
3
581
NM_001326333.1
NP_001313262.1
3
581
NM_001326334.1
NP_001313263.1
4
766
NM_001326335.1
NP_001313264.1
3
581
NM_001326336.1
NP_001313265.1
3
581
NM_001326337.1
NP_001313266.1
3
581
NM_001326340.1
NP_001313269.1
3
493
NM_001326341.1
NP_001313270.1
3
493
NM_001326342.1
NP_001313271.1
3
493
NM_001326343.1
NP_001313272.1
3
493
NM_001326344.1
NP_001313273.1
3
283
NM_001326345.1
NP_001313274.1
3
361
NM_001326347.1
NP_001313276.1
3
691
NM_001326348.1
NP_001313277.1
3
689
NM_001326349.1
NP_001313278.1
3
689
NM_006561.3
NP_006552.3
3
486
XM_005252354.4
XP_005252411.1
3
576
XM_011519294.2
XP_011517596.1
3
576
XM_011519297.1
XP_011517599.1
3
699
XM_017015542.1
XP_016871031.1
2
252
XM_017015543.1
XP_016871032.1
3
342
XM_017015544.1
XP_016871033.1
3
342
XM_017015545.1
XP_016871034.1
3
342
XM_017015546.1
XP_016871035.1
3
342
XM_017015547.1
XP_016871036.1
3
576
XM_017015548.1
XP_016871037.1
3
342
XM_017015549.1
XP_016871038.1
3
342
XM_017015550.1
XP_016871039.1
3
1322
XM_017015551.1
XP_016871040.1
3
576
XM_017015552.1
XP_016871041.1
3
699
XM_017015553.1
XP_016871042.1
3
699
XM_017015554.1
XP_016871043.1
3
699
XM_017015555.1
XP_016871044.1
2
252
XM_017015564.1
XP_016871053.1
2
272
XM_017015566.1
XP_016871055.1
2
272
XM_017015567.1
XP_016871056.1
3
412
XM_017015568.1
XP_016871057.1
5
399
XM_017015569.1
XP_016871058.1
3
699
XM_017015571.1
XP_016871060.1
3
699
XM_017015572.1
XP_016871061.1
3
400
GenBank mRNA
AF036956.1
-
3
456
AF090693.1
-
3
372
AF090694.1
-
3
324
AF432906.1
-
3
392
AK293164.1
-
3
433
AK299548.1
-
3
316
AK299983.1
-
3
605
AK299992.1
-
3
473
AK304216.1
-
2
282
AK314087.1
-
3
495
AK316354.1
-
3
292
BC036391.1
-
3
458
5260623
BP229421.1
-
3
397
BP362205.1
-
1
39
BQ213648.1
-
3
541
DA819821.1
-
2
210
U69546.1
-
3
273
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
regulation of heart contraction
nucleotide binding
RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-zp8kv:80/10.239.163.191:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0