Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206942.1 | NP_001193871.1 | 2 | 1419 | ||
NM_001206943.1 | NP_001193872.1 | 2 | 1341 | |||
NM_001206944.1 | NP_001193873.1 | 2 | 1341 | |||
NM_001206945.1 | NP_001193874.1 | 2 | 1419 | |||
NM_006574.3 | NP_006565.2 | 2 | 1341 | |||
GenBank mRNA | AF059274.1 | - | 2 | 1171 | ||
AF461087.1 | - | 2 | 3264 | |||
AF461088.1 | - | 1 | 1273 | |||
AF461089.1 | - | 2 | 3264 | |||
AK294621.1 | - | 2 | 1419 |