Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319075.1 | NP_001306004.1 | 2 | 688 | ||
NM_006577.5 | NP_006568.2 | 2 | 822 | |||
GenBank mRNA | AB049584.1 | - | 2 | 817 | ||
AF092051.2 | - | 2 | 819 | |||
AF288208.1 | - | 2 | 812 | |||
AF288209.1 | - | 1 | 614 | |||
AJ006077.1 | - | 1 | 587 | |||
AK002009.1 | - | 1 | 517 | |||
BC030579.2 | - | 2 | 691 | 4828158 | ||
BC047933.1 | - | 2 | 822 | 5767816 |