Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319674.1 | NP_001306603.1 | 10 | 1588 | ||
NM_006582.3 | NP_006573.2 | 10 | 1618 | |||
NM_024482.2 | NP_077808.1 | 10 | 1585 | |||
XM_011540518.2 | XP_011538820.1 | 10 | 1721 | |||
XM_011540519.2 | XP_011538821.1 | 10 | 1561 | |||
XM_011540521.2 | XP_011538823.1 | 10 | 1690 | |||
XM_011540522.2 | XP_011538824.1 | 10 | 1537 | |||
XM_017000087.1 | XP_016855576.1 | 10 | 1736 | |||
GenBank mRNA | AF099013.1 | - | 9 | 1482 | ||
AF173868.1 | - | 10 | 1513 | |||
AF203694.1 | - | 10 | 1512 | |||
AK000892.1 | - | 10 | 1584 | |||
AK023126.1 | - | 10 | 1505 | |||
AK315195.1 | - | 10 | 1564 | |||
AK315436.1 | - | 10 | 1522 | |||
BC001473.2 | - | 10 | 1535 | 3138481 |