Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199420.3 | NP_955452.3 | 16 | 4312 | ||
XM_011512343.2 | XP_011510645.1 | 16 | 4300 | |||
XM_011512347.2 | XP_011510649.1 | 16 | 4300 | |||
XM_011512348.2 | XP_011510650.1 | 15 | 4169 | |||
XM_011512352.2 | XP_011510654.1 | 16 | 4300 | |||
XM_011512354.2 | XP_011510656.1 | 4 | 2058 | |||
XM_017005565.1 | XP_016861054.1 | 10 | 3164 | |||
GenBank mRNA | AB209867.1 | - | 16 | 4239 | ||
AF052573.1 | - | 1 | 2112 | |||
AY032677.1 | - | 17 | 4585 | |||
AY338826.2 | - | 16 | 4312 | |||
CR936627.1 | - | 15 | 6362 |