Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016272.3 | NP_057356.1 | 2 | 1261 | ||
XM_005261315.2 | XP_005261372.1 | 2 | 2472 | |||
XM_006724105.3 | XP_006724168.1 | 2 | 1778 | |||
XM_017028539.1 | XP_016884028.1 | 2 | 3554 | |||
GenBank mRNA | AB035207.1 | - | 1 | 768 | ||
AB051450.1 | - | 2 | 1020 | |||
BC028919.1 | - | 2 | 1010 | 4940977 | ||
BC038957.1 | - | 2 | 1034 | 5786640 | ||
CR456594.1 | - | 2 | 714 | |||
CR541684.1 | - | 1 | 554 | |||
D64109.1 | - | 1 | 647 |