Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 10 | 1146 | ||
NM_001166167.1 | NP_001159639.1 | 9 | 928 | |||
NM_001166168.1 | NP_001159640.1 | 9 | 923 | |||
NM_001166169.1 | NP_001159641.1 | 9 | 932 | |||
NM_001166170.1 | NP_001159642.1 | 9 | 930 | |||
NM_001166171.1 | NP_001159643.1 | 10 | 1095 | |||
NM_014397.5 | NP_055212.2 | 9 | 981 | |||
XM_005251664.1 | XP_005251721.1 | 9 | 987 | |||
XM_006716936.2 | XP_006716999.1 | 9 | 992 | |||
XM_017014217.1 | XP_016869706.1 | 10 | 1155 | |||
GenBank mRNA | AB026289.1 | - | 9 | 912 | ||
AF087909.1 | - | 9 | 917 | |||
AK294614.1 | - | 9 | 932 | |||
AK313071.1 | - | 9 | 882 | |||
BC000101.2 | - | 9 | 927 | 3507262 | ||
BC004174.2 | - | 10 | 1099 | 2958695 | ||
BC004209.2 | - | 10 | 1099 | 2958695 | ||
BC012761.2 | - | 9 | 928 | 3640103 | ||
CR457091.1 | - | 8 | 762 | |||
CR542222.1 | - | 8 | 762 |