Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006645.2 | NP_006636.2 | 7 | 1628 | ||
GenBank mRNA | AF039696.1 | - | 251 | |||
AF151810.1 | - | 7 | 1164 | |||
AK095668.1 | - | 7 | 1544 | |||
AK315255.1 | - | 8 | 1422 | |||
BC007919.2 | - | 8 | 1147 | 4301295 | ||
BC014033.2 | - | 8 | 1147 | 4330790 | ||
BC035482.1 | - | 7 | 1428 | |||
JX424571.1 | - | 3 | 133 |