Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGRMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282621.1 | NP_001269550.1 | 1 | 422 | ||
NM_006667.4 | NP_006658.1 | 1 | 422 | |||
GenBank mRNA | AK293618.1 | - | 1 | 342 | ||
AK313956.1 | - | 1 | 380 | |||
BC034238.1 | - | 1 | 356 | 3863377 | ||
CR456993.1 | - | 1 | 303 | |||
CX762728.1 | - | 1 | 419 | |||
HY166171.1 | - | 1 | 422 | |||
Y12711.2 | - | 1 | 381 |