Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014811.1 | NP_001014811.1 | 14 | 2020 | ||
NM_001161586.1 | NP_001155058.1 | 15 | 2093 | |||
NM_006680.2 | NP_006671.2 | 15 | 1868 | |||
XM_005273717.1 | XP_005273774.1 | 15 | 1969 | |||
XM_017017136.1 | XP_016872625.1 | 15 | 2851 | |||
XM_017017137.1 | XP_016872626.1 | 11 | 1393 | |||
XM_017017138.1 | XP_016872627.1 | 16 | 1977 | |||
GenBank mRNA | AI198184.1 | - | 219 | |||
AI560417.1 | - | 1 | 215 | |||
AI948779.1 | - | 1 | 217 | |||
AK313695.1 | - | 15 | 1943 | |||
BC022472.2 | - | 14 | 2020 | 4812045 | ||
BQ882896.1 | - | 6 | 754 | |||
CB306059.1 | - | 1 | 233 | |||
X79440.1 | - | 14 | 1798 |