Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014599.5 | NP_055414.2 | 4, 5 | 981 | ||
NM_177433.2 | NP_803182.1 | 4, 5 | 1089 | |||
NM_201222.2 | NP_957516.1 | 4, 5 | 1049 | |||
GenBank mRNA | AF126181.1 | - | 4, 5 | 925 | ||
AF128527.1 | - | 4, 5 | 925 | |||
AF128528.1 | - | 4, 5 | 925 | |||
AJ293618.1 | - | 3, 4 | 868 | |||
AK092463.1 | - | 5, 6 | 887 | |||
AK098645.1 | - | 6, 7 | 693 | |||
AK297910.1 | - | 4, 5 | 696 | |||
AK301599.1 | - | 4, 5 | 684 | |||
AK301821.1 | - | 4, 5 | 666 | |||
BC000304.2 | - | 4, 5 | 943 | 2820442 | ||
BC091503.1 | - | 4, 5 | 921 | 30529545 | ||
BX647995.1 | - | 4, 5 | 1026 | |||
U92544.1 | - | 4, 5 | 922 |