Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006805.3 | NP_006796.1 | 1 | 673 | ||
GenBank mRNA | BC001008.1 | - | 1 | 672 | 2964662 | |
BC007271.1 | - | 1 | 401 | 3160840 | ||
BC009284.2 | - | 1 | 445 | 4299262 | ||
BC011972.1 | - | 1 | 664 | 3454140 | ||
BC012980.2 | - | 1 | 672 | 3617551 | ||
BC018800.1 | - | 1 | 670 | |||
BC018949.2 | - | 1 | 672 | 4131137 | ||
BC019271.1 | - | 1 | 652 | 2823669 | ||
BC028976.1 | - | 1 | 673 | 4993025 | ||
BC030249.1 | - | 1 | 673 | 5242912 | ||
CR456986.1 | - | 1 | 389 | |||
L10372.1 | - | 1 | 76 | |||
U23803.1 | - | 1 | 653 |