Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006821.5 | NP_006812.3 | 2, 3 | 1019 | ||
NR_046028.1 | 3, 4 | 823 | ||||
XM_006720007.1 | XP_006720070.1 | 2, 3 | 506 | |||
GenBank mRNA | AK001939.1 | - | 2, 3 | 1019 | ||
AK093131.1 | - | 3, 4 | 823 | |||
AK223635.1 | - | 2, 3 | 1015 | |||
AY005822.1 | - | 2, 3 | 735 | |||
BC004436.1 | - | 2, 3 | 793 | |||
BC006335.2 | - | 2, 3 | 1003 | 4303771 | ||
BC006500.2 | - | 2, 3 | 793 | |||
DQ082755.1 | - | 2, 3 | 1019 | |||
L40401.1 | - | 2, 3 | 322 |