Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 15 | 2377 | ||
NM_001100170.1 | NP_001093640.1 | 15 | 2347 | |||
NM_006839.2 | NP_006830.2 | 15 | 2380 | |||
XM_005264110.3 | XP_005264167.1 | 15 | 2016 | |||
XM_005264113.1 | XP_005264170.1 | 14 | 1952 | |||
XM_005264114.1 | XP_005264171.1 | 14 | 1923 | |||
XM_011532497.2 | XP_011530799.1 | 15 | 2040 | |||
XM_017003194.1 | XP_016858683.1 | 15 | 2048 | |||
XM_017003195.1 | XP_016858684.1 | 15 | 2072 | |||
GenBank mRNA | AF148646.1 | - | 10 | 1548 | ||
AK225780.1 | - | 15 | 2014 | |||
AK296676.1 | - | 15 | 1986 | |||
AK299014.1 | - | 14 | 1902 | |||
AK299596.1 | - | 13 | 1757 | |||
AK300017.1 | - | 13 | 1723 | |||
AK312251.1 | - | 15 | 2030 | |||
AY232292.1 | - | 15 | 2010 | |||
AY921637.1 | - | 15 | 2028 | |||
BC002412.2 | - | 15 | 2021 | 2964676 | ||
BC027458.1 | - | 4 | 733 | |||
BC107419.1 | - | 8 | 918 | |||
BX537369.1 | - | 15 | 2051 | |||
CR456793.1 | - | 15 | 1992 | |||
CR749590.1 | - | 10 | 1424 | |||
D21092.1 | - | 13 | 1882 | |||
D21094.1 | - | 15 | 2077 | |||
L42572.1 | - | 15 | 2084 |