Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDELR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006855.3 | NP_006846.1 | 3 | 467 | ||
NM_016657.2 | NP_057839.1 | 3 | 467 | |||
GenBank mRNA | AK129888.1 | - | 3 | 433 | ||
AK222930.1 | - | 3 | 447 | |||
AK223238.1 | - | 3 | 424 | |||
AK292102.1 | - | 3 | 459 | |||
AL035081.1 | - | 3 | 420 | |||
AL035082.1 | - | 3 | 451 | |||
BC001277.1 | - | 3 | 444 | 3462392 | ||
BC096722.1 | - | 3 | 401 | 40003712 | ||
BC098131.1 | - | 3 | 401 | 40003710 | ||
BC098156.1 | - | 3 | 401 | 40003711 | ||
BC098336.1 | - | 3 | 401 | 40003709 | ||
BU621350.1 | - | 2 | 490 | |||
CR456509.1 | - | 3 | 342 |