Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100397.1 | NP_001093867.1 | 18, 19 | 1950 | ||
NM_001282899.1 | NP_001269828.1 | 18, 19 | 1821 | |||
NM_001282900.1 | NP_001269829.1 | 17, 18 | 1767 | |||
NM_001282901.1 | NP_001269830.1 | 15, 16 | 1555 | |||
NM_007023.3 | NP_008954.2 | 21, 22 | 2191 | |||
XM_005246246.2 | XP_005246303.1 | 20, 21 | 2004 | |||
XM_006712205.3 | XP_006712268.1 | 20, 21 | 2009 | |||
XM_011510505.2 | XP_011508807.1 | 20, 21 | 2038 | |||
XM_017003196.1 | XP_016858685.1 | 21, 22 | 2058 | |||
XM_017003197.1 | XP_016858686.1 | 17, 18 | 1707 | |||
GenBank mRNA | AB027471.1 | - | 21, 22 | 2061 | ||
AB075513.1 | - | 5, 6 | 406 | |||
AB209681.1 | - | 21, 22 | 4245 | |||
AK294433.1 | - | 15, 16 | 1479 | |||
AK294445.1 | - | 18, 19 | 1670 | |||
AK294914.1 | - | 15, 16 | 1508 | |||
AK295002.1 | - | 17, 18 | 1680 | |||
AK296340.1 | - | 17, 18 | 1762 | |||
AK304278.1 | - | 18, 19 | 1710 | |||
AK313084.1 | - | 21, 22 | 2060 | |||
AK316014.1 | - | 18, 19 | 1785 | |||
AK316072.1 | - | 18, 19 | 1717 | |||
BC024004.2 | - | 21, 22 | 2030 | 4823935 | ||
BC040183.2 | - | 18, 19 | 1951 | 4801092 | ||
KJ534924.1 | - | 21, 22 | 1968 | |||
KJ534925.1 | - | 20, 21 | 1950 | |||
KJ534978.1 | - | 21, 22 | 2004 | |||
U78516.1 | - | 21, 22 | 2112 |