Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300791.1 | NP_001287720.1 | 2 | 204 | ||
NM_001300792.1 | NP_001287721.1 | 2 | 204 | |||
NM_007054.6 | NP_008985.3 | 2 | 204 | |||
XM_006714526.3 | XP_006714589.1 | 2 | 259 | |||
XM_017008995.1 | XP_016864484.1 | 2 | 259 | |||
XM_017008996.1 | XP_016864485.1 | 2 | 259 | |||
GenBank mRNA | AB209780.1 | - | 2 | 152 | ||
AF041853.1 | - | 1 | 45 | |||
AK295089.1 | - | 2 | 162 | |||
AK313359.1 | - | 2 | 172 | |||
AM177178.1 | - | 1 | 45 | |||
BC032599.1 | - | 2 | 187 | |||
BC039592.1 | - | 2 | 161 | |||
BC045542.1 | - | 2 | 198 | 5298675 | ||
BI561509.1 | - | 2 | 204 | |||
HY154618.1 | - | 2 | 204 |