Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007065.3 | NP_008996.1 | 6, 7 | 1032 | ||
XM_011527652.2 | XP_011525954.1 | 6, 7 | 1152 | |||
GenBank mRNA | BC000083.2 | - | 6, 7 | 956 | 3505011 | |
BC008793.2 | - | 6, 7 | 956 | 3619769 | ||
BC127212.1 | - | 1, 2 | 112 | |||
BC127213.1 | - | 1, 2 | 112 | |||
BC141928.1 | - | 1, 2 | 112 | |||
BC141929.1 | - | 1, 2 | 112 | |||
BT006796.1 | - | 6, 7 | 916 | |||
CR542249.1 | - | 6, 7 | 916 | |||
U43077.1 | - | 6, 7 | 924 | |||
U63131.1 | - | 6, 7 | 964 |