Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301065.1 | NP_001287994.1 | 12 | 1445 | ||
NM_007180.2 | NP_009111.2 | 13 | 1538 | |||
GenBank mRNA | AB000824.1 | - | 13 | 1548 | ||
AK091981.1 | - | 11 | 1407 | |||
AK223140.1 | - | 13 | 1512 | |||
AK223143.1 | - | 13 | 1516 | |||
BC109206.2 | - | 12 | 1439 | 40006501 |