Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007217.3 | NP_009148.2 | 6 | 694 | ||
NM_145859.1 | NP_665858.1 | 6 | 543 | |||
NM_145860.1 | NP_665859.1 | 5 | 449 | |||
XM_005247086.4 | XP_005247143.1 | 6 | 1429 | |||
XM_005247087.4 | XP_005247144.1 | 6 | 1434 | |||
XM_005247088.3 | XP_005247145.1 | 5 | 525 | |||
XM_006713485.3 | XP_006713548.1 | 6 | 589 | |||
XM_011512368.2 | XP_011510670.1 | 7 | 1502 | |||
XM_011512369.2 | XP_011510671.1 | 7 | 1507 | |||
XM_017005644.1 | XP_016861133.1 | 6 | 584 | |||
XM_017005645.1 | XP_016861134.1 | 6 | 1425 | |||
GenBank mRNA | AK291130.1 | - | 5 | 480 | ||
BC002506.1 | - | 6 | 706 | 3050540 | ||
BC016353.1 | - | 6 | 547 | 4092244 | ||
CR457107.1 | - | 4 | 296 |