Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016219.4 | NP_057303.2 | 13 | 2068 | ||
NR_045720.1 | 13 | 2043 | ||||
NR_045721.1 | 14 | 2199 | ||||
XM_006716945.3 | XP_006717008.1 | 12 | 2858 | |||
XM_017014239.1 | XP_016869728.1 | 13 | 2770 | |||
GenBank mRNA | AB209276.1 | - | 4 | 2788 | ||
AF145732.1 | - | 13 | 1985 | |||
AF148509.1 | - | 13 | 2045 | |||
AK074559.1 | - | 5 | 632 | |||
AK074699.1 | - | 11 | 1643 | |||
AK094878.1 | - | 5 | 2074 | |||
AK126926.1 | - | 6 | 4102 | |||
AK127005.1 | - | 6 | 3167 | |||
AK128033.1 | - | 5 | 4353 | |||
AK128257.1 | - | 6 | 3245 | |||
AK128341.1 | - | 6 | 3915 | |||
AK130143.1 | - | 8 | 1212 | |||
AK298480.1 | - | 14 | 2003 | |||
AK315797.1 | - | 13 | 1985 | |||
AL110221.1 | - | 5 | 1515 | |||
AY358465.1 | - | 13 | 2017 | |||
BC002953.1 | - | 13 | 2001 | 3533651 | ||
BC006079.1 | - | 13 | 2010 | 3959196 | ||
CR749534.1 | - | 19 | 6551 |