Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317192.1 | NP_001304121.1 | 1 | 147 | ||
NM_001317193.1 | NP_001304122.1 | 1 | 147 | |||
NM_001317194.1 | NP_001304123.1 | 1 | 147 | |||
NM_007241.3 | NP_009172.2 | 1 | 147 | |||
NR_133679.1 | 1 | 147 | ||||
XM_017024075.1 | XP_016879564.1 | 1 | 89 | |||
GenBank mRNA | AF156102.1 | - | 1 | 96 | ||
BC008976.1 | - | 1 | 97 | 3452153 | ||
BC038830.1 | - | 1 | 121 | 3900806 | ||
BG248541.1 | - | 1 | 136 | |||
BP342490.1 | - | 1 | 147 |