Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135752.1 | NP_001129224.1 | 11 | 1496 | ||
NM_001135753.1 | NP_001129225.1 | 9 | 1268 | |||
NM_007265.2 | NP_009196.1 | 10 | 1397 | |||
NR_024203.1 | 8 | 986 | ||||
GenBank mRNA | AK022616.1 | - | 10 | 1345 | ||
AK225519.1 | - | 9 | 1244 | |||
AK315711.1 | - | 11 | 1469 | |||
BC000721.2 | - | 10 | 1361 | 3504956 | ||
CR457407.1 | - | 9 | 1154 | |||
D88208.1 | - | 10 | 1361 |